4-(4-Fluorophenyl)-3-methyl-1H-1,2,4-triazol-5-one

Lieferant: Apollo Scientific
Achtung

PC32895-1G PC32895-250MG
APOSPC32895-1GEA 183 EUR
APOSPC32895-1G APOSPC32895-250MG
4-(4-Fluorophenyl)-3-methyl-1H-1,2,4-triazol-5-one
4-(4-Fluorophenyl)-3-methyl-1H-1,2,4-triazol-5-one
Formel: C₉H₈FN₃O
Molecular Weight: 193,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787254
CAS-Nummer: 860650-96-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR