2-Fluoro-4-methyl-5-nitrobenzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
PC32910-1G
PC32910-250MG
APOSPC32910-1GEA
174
EUR
APOSPC32910-1G
APOSPC32910-250MG
2-Fluoro-4-methyl-5-nitrobenzyl alcohol
2-Fluoro-4-methyl-5-nitrobenzyl alcohol
Formel:
C₈H₈FNO₃ Molecular Weight: 185,152 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28014049 CAS-Nummer: 1806480-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...