2-Fluoro-4-methyl-5-nitrobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC32911-1G PC32911-250MG PC32911-5G
APOSPC32911-1GEA 464 EUR
APOSPC32911-1G APOSPC32911-250MG APOSPC32911-5G
2-Fluoro-4-methyl-5-nitrobenzyl bromide
2-Fluoro-4-methyl-5-nitrobenzyl bromide
Formel: C₈H₇BrFNO₂
Molecular Weight: 248,049 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28797798
CAS-Nummer: 1803833-24-0
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR