4-Bromo-2-[bromo(difluoro)methyl]-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

PC32917-1G PC32917-250MG PC32917-5G
APOSPC32917-1GEA 140 EUR
APOSPC32917-1G APOSPC32917-250MG APOSPC32917-5G
4-Bromo-2-[bromo(difluoro)methyl]-1H-benzimidazole
4-Bromo-2-[bromo(difluoro)methyl]-1H-benzimidazole
Formel: C₈H₄Br₂F₂N₂
Molecular Weight: 325,936 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31922500
CAS-Nummer: 2366994-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR