2-[Bromo(difluoro)methyl]-5-fluoro-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

PC32920-1G PC32920-250MG PC32920-5G
APOSPC32920-1GEA 48 EUR
APOSPC32920-1G APOSPC32920-250MG APOSPC32920-5G
2-[Bromo(difluoro)methyl]-5-fluoro-1H-benzimidazole
2-[Bromo(difluoro)methyl]-5-fluoro-1H-benzimidazole
Formel: C₈H₄BrF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31922503
CAS-Nummer: 2366994-42-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR