2-[Bromo(difluoro)methyl]-5-(trifluoromethyl)-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

PC32924-1G PC32924-250MG PC32924-5G
APOSPC32924-1GEA 109 EUR
APOSPC32924-1G APOSPC32924-250MG APOSPC32924-5G
2-[Bromo(difluoro)methyl]-5-(trifluoromethyl)-1H-benzimidazole
2-[Bromo(difluoro)methyl]-5-(trifluoromethyl)-1H-benzimidazole
Formel: C₉H₄BrF₅N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31922507
CAS-Nummer: 1353778-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR