2-(Difluoromethyl)-4-nitro-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

PC32938-1G PC32938-250MG PC32938-5G
APOSPC32938-1GEA 208 EUR
APOSPC32938-1G APOSPC32938-250MG APOSPC32938-5G
2-(Difluoromethyl)-4-nitro-1H-benzimidazole
2-(Difluoromethyl)-4-nitro-1H-benzimidazole
Formel: C₈H₅F₂N₃O₂
Molecular Weight: 213,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22561278
CAS-Nummer: 189045-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR