5-Fluoro-7-methoxy-4(3H)-quinazolinone

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-7-methoxyquinazolin-4(3H)-one

PC32957-100MG PC32957-1G PC32957-250MG
APOSPC32957-100MGEA 87 EUR
APOSPC32957-100MG APOSPC32957-1G APOSPC32957-250MG
5-Fluoro-7-methoxy-4(3H)-quinazolinone
5-Fluoro-7-methoxy-4(3H)-quinazolinone
Formel: C₉H₇FN₂O₂
Molecular Weight: 194,162 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD13191851
CAS-Nummer: 944742-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR