5-(Difluoromethoxy)picolinic acid 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Difluoromethoxy)pyridine-2-carboxylic acid

PC32959-1G PC32959-250MG
APOSPC32959-1GEA 424 EUR
APOSPC32959-1G APOSPC32959-250MG
5-(Difluoromethoxy)picolinic acid 99%
5-(Difluoromethoxy)picolinic acid
Formel: C₇H₅F₂NO₃
Molecular Weight: 189.116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18261162
CAS-Nummer: 1174323-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR