Methyl 5-(4-(trifluoromethoxy)phenyl)thiophene-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC32963-1G
PC32963-250MG
APOSPC32963-1GEA
140
EUR
APOSPC32963-1G
APOSPC32963-250MG
Methyl 5-(4-(trifluoromethoxy)phenyl)thiophene-2-carboxylate
Methyl 5-(4-(trifluoromethoxy)phenyl)thiophene-2-carboxylate
Formel:
C₁₃H₉F₃O₃S Molecular Weight: 302,269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18408646 CAS-Nummer: 1261530-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...