3-Amino-6-(4-fluorophenyl)pyridazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(4-Fluorophenyl)pyridazin-3-amine, 1-(6-Aminopyridazin-3-yl)-4-fluorobenzene

PC3314-100MG PC3314-250MG
APOSPC3314-100MGEA 210 EUR
APOSPC3314-100MG APOSPC3314-250MG
3-Amino-6-(4-fluorophenyl)pyridazine
3-Amino-6-(4-fluorophenyl)pyridazine
Formel: C₁₀H₈FN₃
Molecular Weight: 189,189 g/mol
Schmelzpunkt: 170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06801351
CAS-Nummer: 105538-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR