2,3-Difluoro-1-iodo-4-(trifluoromethoxy)benzene 95%

Lieferant: Apollo Scientific
Achtung

PC33176-1G PC33176-250MG PC33176-5G
APOSPC33176-1GEA 145 EUR
APOSPC33176-1G APOSPC33176-250MG APOSPC33176-5G
2,3-Difluoro-1-iodo-4-(trifluoromethoxy)benzene 95%
2,3-Difluoro-1-iodo-4-(trifluoromethoxy)benzene
Formel: C₇H₂F₅IO
Molecular Weight: 323.987 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32066740
CAS-Nummer: 2366994-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR