2-Fluoro-5-(4-methoxyphenyl)pyridine

Lieferant: Apollo Scientific

PC3361-1G PC3361-250MG
APOSPC3361-1GEA 198 EUR
APOSPC3361-1G APOSPC3361-250MG
2-Fluoro-5-(4-methoxyphenyl)pyridine
2-Fluoro-5-(4-methoxyphenyl)pyridine
Formel: C₁₂H₁₀FNO
Molecular Weight: 203,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907672
CAS-Nummer: 444120-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR