2-(Difluoromethoxy)phenol 98%
Lieferant: Apollo Scientific
PC3421-1G
PC3421-250MG
PC3421-500MG
PC3421-5G
APOSPC3421-1GEA
71.2
EUR
APOSPC3421-1G
APOSPC3421-250MG
APOSPC3421-500MG
APOSPC3421-5G
2-(Difluoromethoxy)phenol 98%
2-(Difluoromethoxy)phenol
Formel:
C₇H₆F₂O₂ Molecular Weight: 160,118 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236234 CAS-Nummer: 53104-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...