1,2,3,6-Tetrahydro-4-(trifluoromethyl)pyridine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

PC3461-1G PC3461-250MG
APOSPC3461-1GEA 673 EUR
APOSPC3461-1G APOSPC3461-250MG
1,2,3,6-Tetrahydro-4-(trifluoromethyl)pyridine hydrochloride 95%
1,2,3,6-Tetrahydro-4-(trifluoromethyl)pyridine hydrochloride
Formel: C₆H₈F₃N·ClH
Molecular Weight: 187.591 g/mol
Schmelzpunkt: 95…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11035860
CAS-Nummer: 266359-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR