4-Fluoro-5-hydroxy-1H-indole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-1H-indol-5-ol
PC3466-1G
PC3466-250MG
APOSPC3466-1GEA
603
EUR
APOSPC3466-1G
APOSPC3466-250MG
4-Fluoro-5-hydroxy-1H-indole
4-Fluoro-5-hydroxy-1H-indole
Formel:
C₈H₆FNO Molecular Weight: 151,138 g/mol Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07368139 CAS-Nummer: 288386-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...