2,3-Difluoro-4-(trifluoromethyl)benzyl bromide 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(Bromomethyl)-2,3-difluoro-4-(trifluoromethyl)benzene, 4-(Bromomethyl)-2,3-difluorobenzotrifluoride

PC3475-1G PC3475-250MG
APOSPC3475-1GEA 279 EUR
APOSPC3475-1G APOSPC3475-250MG
2,3-Difluoro-4-(trifluoromethyl)benzyl bromide 98%
2,3-Difluoro-4-(trifluoromethyl)benzyl bromide
Formel: C₈H₄BrF₅
Molecular Weight: 275.013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236262
CAS-Nummer: 213203-85-1
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR