1-[3-(Trifluoromethyl)phenyl]propylamine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[3-(Trifluoromethyl)phenyl]propan-1-amine

PC3491-1G PC3491-250MG
APOSPC3491-1GEA 321 EUR
APOSPC3491-1G APOSPC3491-250MG
1-[3-(Trifluoromethyl)phenyl]propylamine 98%
1-[3-(Trifluoromethyl)phenyl]propylamine
Formel: C₁₀H₁₂F₃N
Molecular Weight: 203,204 g/mol
Flammpunkt: 90 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07784289
CAS-Nummer: 473732-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR