2-Fluoro-4-nitrophenyl methyl sulphone

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-4-(methylsulphonyl)nitrobenzene, 2-Fluoro-1-(methylsulphonyl)-4-nitrobenzene

PC3502-1G PC3502-250MG
APOSPC3502-1GEA 62.5 EUR
APOSPC3502-1G APOSPC3502-250MG
2-Fluoro-4-nitrophenyl methyl sulphone
2-Fluoro-4-nitrophenyl methyl sulphone
Formel: C₇H₆FNO₄S
Molecular Weight: 219.19 g/mol
Schmelzpunkt: 101…103 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08692411
CAS-Nummer: 252561-33-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR