Bis(3,3,3-trifluoroprop-1-yl)amine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3,3,3-Trifluoro-N-(3,3,3-trifluoroprop-1-yl)propylamine

PC3539-1G PC3539-5G
APOSPC3539-1GEA 180 EUR
APOSPC3539-1G APOSPC3539-5G
Bis(3,3,3-trifluoroprop-1-yl)amine 95%
Bis(3,3,3-trifluoroprop-1-yl)amine
Formel: C₆H₉F₆N
Molecular Weight: 209,133 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07784275
CAS-Nummer: 201057-04-7
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR