2,3-Dichloro-6-fluorophenacyl bromide

Lieferant: Apollo Scientific
Gefahr

PC3541-10G PC3541-5G
APOSPC3541-10GEA 445 EUR
APOSPC3541-10G APOSPC3541-5G
2,3-Dichloro-6-fluorophenacyl bromide
2,3-Dichloro-6-fluorophenacyl bromide
Formel: C₈H₄BrCl₂FO
Molecular Weight: 285,925 g/mol
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368755
CAS-Nummer: 886762-53-4
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR