2-[(2-Fluorophenyl)amino]nitrobenzene 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-2'-nitrodiphenylamine, 2-Fluoro-N-(2-nitrophenyl)aniline
PC3545-25G
PC3545-5G
APOSPC3545-25GEA
277
EUR
APOSPC3545-25G
APOSPC3545-5G
2-[(2-Fluorophenyl)amino]nitrobenzene 98%
2-[(2-Fluorophenyl)amino]nitrobenzene
Formel:
C₁₂H₉FN₂O₂ Molecular Weight: 232,21 g/mol Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08741396 CAS-Nummer: 28898-02-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...