2-Fluoro-5-(hydrazinocarbonyl)benzeneboronic acid 95%
Lieferant: Apollo Scientific
Synonyme:
3-Borono-4-fluorobenzohydrazide, 2-Fluoro-5-(hydrazinecarbonyl)phenylboronic acid
PC3561-1G
PC3561-5G
APOSPC3561-1GEA
345
EUR
APOSPC3561-1G
APOSPC3561-5G
2-Fluoro-5-(hydrazinocarbonyl)benzeneboronic acid 95%
2-Fluoro-5-(hydrazinocarbonyl)benzeneboronic acid
Formel:
C₇H₈BFN₂O₃ Molecular Weight: 197,959 g/mol Schmelzpunkt: 183…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08436072 CAS-Nummer: 874289-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...