3-Bromo-5-fluorobenzene-1,2-diamine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-2,3-diamino-5-fluorobenzene, 3-Bromo-5-fluorophenylene-1,2-diamine

PC3592-1G PC3592-5G PC3592-25G
APOSPC3592-1GEA 23.2 EUR
APOSPC3592-1G APOSPC3592-5G APOSPC3592-25G
3-Bromo-5-fluorobenzene-1,2-diamine 97%
3-Bromo-5-fluorobenzene-1,2-diamine
Formel: C₆H₆BrFN₂
Molecular Weight: 205,028 g/mol
Schmelzpunkt: 70,3…70.6 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08741398
CAS-Nummer: 115440-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR