2-Fluoro-4-(2-methylallyl)chlorobenzene

Lieferant: Apollo Scientific

Synonyme: 3-(4-Chloro-3-fluorophenyl)-2-methylprop-1-ene

PC3641-1G PC3641-5G
APOSPC3641-1GEA 239 EUR
APOSPC3641-1G APOSPC3641-5G
2-Fluoro-4-(2-methylallyl)chlorobenzene
2-Fluoro-4-(2-methylallyl)chlorobenzene
Formel: C₁₀H₁₀ClF
Molecular Weight: 184,638 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774995
CAS-Nummer: 787585-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR