2-Fluoro-4-(2-methylallyl)bromobenzene
Lieferant: Apollo Scientific
Synonyme:
3-(4-Bromo-3-fluorophenyl)-2-methylprop-1-ene
PC3645-1G
PC3645-5G
APOSPC3645-1GEA
239
EUR
APOSPC3645-1G
APOSPC3645-5G
2-Fluoro-4-(2-methylallyl)bromobenzene
2-Fluoro-4-(2-methylallyl)bromobenzene
Formel:
C₁₀H₁₀BrF Molecular Weight: 229,089 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09801086 CAS-Nummer: 951888-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...