Nonafluorobutanesulphonic acid hydrate 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Perfluorobutanesulphonic acid hydrate

PC3681-25G PC3681-5G
APOSPC3681-25GEA 317 EUR
APOSPC3681-25G APOSPC3681-5G
Nonafluorobutanesulphonic acid hydrate 95%
Nonafluorobutanesulphonic acid hydrate
Formel: C₄HF₉O₃S·H₂O
Molecular Weight: 318,115 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03410279
CAS-Nummer: 1185303-26-7
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR