2-(4-Fluorophenyl)-1H-benzimidazole-5-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Carboxy-2-(4-fluorophenyl)-1H-benzimidazole
PC3747-1G
PC3747-250MG
APOSPC3747-1GEA
82.8
EUR
APOSPC3747-1G
APOSPC3747-250MG
2-(4-Fluorophenyl)-1H-benzimidazole-5-carboxylic acid
2-(4-Fluorophenyl)-1H-benzimidazole-5-carboxylic acid
Formel:
C₁₄H₉FN₂O₂ Molecular Weight: 256,232 g/mol Schmelzpunkt: 304…307 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD10007771 CAS-Nummer: 850705-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...