3-Bromo-4-(trifluoromethoxy)benzene-1-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
PC37867-1G
PC37867-250MG
PC37867-5G
APOSPC37867-1GEA
85.7
EUR
APOSPC37867-1G
APOSPC37867-250MG
APOSPC37867-5G
3-Bromo-4-(trifluoromethoxy)benzene-1-sulphonyl chloride
3-Bromo-4-(trifluoromethoxy)benzene-1-sulphonyl chloride
Formel:
C₇H₃BrClF₃O₃S Molecular Weight: 339,514 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026153 CAS-Nummer: 883146-06-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...