3,5-Dichloro-4-fluorobenzene-1-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

PC37868-1G PC37868-250MG
APOSPC37868-1GEA 212 EUR
APOSPC37868-1G APOSPC37868-250MG
3,5-Dichloro-4-fluorobenzene-1-sulphonyl chloride
3,5-Dichloro-4-fluorobenzene-1-sulphonyl chloride
Formel: C₆H₂Cl₃FO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD12026220
CAS-Nummer: 1131397-79-9
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR