2-(Perfluoroalkyl)ethanol Ultrahigh MW Distribution

Lieferant: Apollo Scientific
Achtung

PC3854-100G PC3854-25G
APOSPC3854-100GEA 256 EUR
APOSPC3854-100G APOSPC3854-25G
2-(Perfluoroalkyl)ethanol Ultrahigh MW Distribution
2-(Perfluoroalkyl)ethanol

Mixture where x = 5,6,7,8.....

Schmelzpunkt: >100 °C
Dichte: 1,7 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00197963
CAS-Nummer: 68391-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR