Pentafluoroallyl fluorosulphate 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1,1,2,3,3-Pentafluoroprop-2-en-1-yl sulphurofluoridoate, Perfluoroallyl fluorosulphate

PC3858-1G PC3858-5G
APOSPC3858-1GEA 219 EUR
APOSPC3858-1G APOSPC3858-5G
Pentafluoroallyl fluorosulphate 97%
Pentafluoroallyl fluorosulphate

Nucleophilic displacement of fluorosulphate anion with iodide affords perfluoroallyl iodide, Krespan & England, J. Amer. Chem. Soc., 103, 5598, 1981

Formel: C₃F₆O₃S
Molecular Weight: 230,086 g/mol
Siedepunkt: 63…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093345
CAS-Nummer: 67641-28-5
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR