2-Amino-4-(4-fluorophenyl)pyrimidine

Lieferant: Apollo Scientific

Synonyme: 4-(4-Fluorophenyl)pyrimidin-2-amine

PC3864-1G PC3864-250MG
APOSPC3864-1GEA 203 EUR
APOSPC3864-1G APOSPC3864-250MG
2-Amino-4-(4-fluorophenyl)pyrimidine
2-Amino-4-(4-fluorophenyl)pyrimidine
Formel: C₁₀H₈FN₃
Molecular Weight: 189.189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407836
CAS-Nummer: 85979-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR