2-Nitro-3,4,5-trifluorobenzoyl chloride

Lieferant: Apollo Scientific
Gefahr

PC3930-10G PC3930-1G PC3930-5G
APOSPC3930-10GEA 621 EUR
APOSPC3930-10G APOSPC3930-1G APOSPC3930-5G
2-Nitro-3,4,5-trifluorobenzoyl chloride
2-Nitro-3,4,5-trifluorobenzoyl chloride
Formel: C₇HClF₃NO₃
Molecular Weight: 239,536 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094231
CAS-Nummer: 173589-92-9
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR