6-Bromo-2,3,4-trifluorobenzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

PC39455-1G PC39455-5G PC39455-25G
APOSPC39455-1GEA 85.7 EUR
APOSPC39455-1G APOSPC39455-5G APOSPC39455-25G
6-Bromo-2,3,4-trifluorobenzaldehyde 97%
6-Bromo-2,3,4-trifluorobenzaldehyde
Formel: C₇H₂BrF₃O
Molecular Weight: 238.989 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15527216
CAS-Nummer: 372519-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR