7-Fluoro-2,3-dihydro-2,2-dimethylchromen-4-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Fluoro-2,2-dimethylchroman-4-one, 7-Fluoro-2,2-dimethyl-3H-chromen-4-one

PC400032-1G PC400032-250MG
APOSPC400032-1GEA 140 EUR
APOSPC400032-1G APOSPC400032-250MG
7-Fluoro-2,3-dihydro-2,2-dimethylchromen-4-one
7-Fluoro-2,3-dihydro-2,2-dimethylchromen-4-one
Formel: C₁₁H₁₁FO₂
Molecular Weight: 194,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15527098
CAS-Nummer: 111477-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR