(2-Bromo-3,4-difluorophenyl)methanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC400037-100MG PC400037-250MG
APOSPC400037-100MGEA 149 EUR
APOSPC400037-100MG APOSPC400037-250MG
(2-Bromo-3,4-difluorophenyl)methanamine hydrochloride
(2-Bromo-3,4-difluorophenyl)methanamine hydrochloride
Formel: C₇H₆BrF₂N·ClH
Molecular Weight: 258,491 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561224
CAS-Nummer: 2231675-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR