5-Bromo-6-(difluoromethoxy)pyridine-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-6-(difluoromethoxy)nicotinonitrile

PC400105-100MG PC400105-250MG
APOSPC400105-100MGEA 179 EUR
APOSPC400105-100MG APOSPC400105-250MG
5-Bromo-6-(difluoromethoxy)pyridine-3-carbonitrile
5-Bromo-6-(difluoromethoxy)pyridine-3-carbonitrile
Formel: C₇H₃BrF₂N₂O
Molecular Weight: 249.012 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28734727
CAS-Nummer: 1805591-98-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR