5-Bromo-6-(difluoromethoxy)pyridine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC400106-100MG PC400106-250MG
APOSPC400106-100MGEA 183 EUR
APOSPC400106-100MG APOSPC400106-250MG
5-Bromo-6-(difluoromethoxy)pyridine-3-carboxylic acid
5-Bromo-6-(difluoromethoxy)pyridine-3-carboxylic acid
Formel: C₇H₄BrF₂NO₃
Molecular Weight: 268.012 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621143
CAS-Nummer: 2231674-35-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR