tert-Butyl (5-bromo-6-(difluoromethoxy)pyridin-3-yl)methylcarbamate

Lieferant: Apollo Scientific
Achtung

PC400109-100MG PC400109-250MG
APOSPC400109-100MGEA 203 EUR
APOSPC400109-100MG APOSPC400109-250MG
tert-Butyl (5-bromo-6-(difluoromethoxy)pyridin-3-yl)methylcarbamate
tert-Butyl (5-bromo-6-(difluoromethoxy)pyridin-3-yl)methylcarbamate
Formel: C₁₂H₁₅BrF₂N₂O₃
Molecular Weight: 353.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621153
CAS-Nummer: 2231674-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR