4-(Difluoromethoxy)-3-fluoropyridine-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC400114-250MG
PC400114-100MG
APOSPC400114-250MGEA
870
EUR
APOSPC400114-250MG
APOSPC400114-100MG
4-(Difluoromethoxy)-3-fluoropyridine-2-carboxylic acid
4-(Difluoromethoxy)-3-fluoropyridine-2-carboxylic acid
Formel:
C₇H₄F₃NO₃ Molecular Weight: 207,107 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28789683 CAS-Nummer: 1807188-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...