1-(5-Bromo-3-fluoropyridin-2-yl)-2,2,2-trifluoroethanone

Lieferant: Apollo Scientific
Achtung

PC400146-1G PC400146-250MG
APOSPC400146-1GEA 0 EUR
APOSPC400146-1G APOSPC400146-250MG
1-(5-Bromo-3-fluoropyridin-2-yl)-2,2,2-trifluoroethanone
1-(5-Bromo-3-fluoropyridin-2-yl)-2,2,2-trifluoroethanone
Formel: C₇H₂BrF₄NO
Molecular Weight: 271,994 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621150
CAS-Nummer: 2231673-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR