2-(Difluoromethoxy)pyridine-3-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Difluoromethoxy)nicotinonitrile
PC400157-250MG
PC400157-100MG
APOSPC400157-250MGEA
115
EUR
APOSPC400157-250MG
APOSPC400157-100MG
2-(Difluoromethoxy)pyridine-3-carbonitrile
2-(Difluoromethoxy)pyridine-3-carbonitrile
Formel:
C₇H₄F₂N₂O Molecular Weight: 170,116 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14698406 CAS-Nummer: 1214388-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...