(3-Chloro-5-fluoropyridin-2-yl)methanamine hydrochloride ≥95%

Lieferant: Apollo Scientific
Achtung

PC400170-1G PC400170-250MG
APOSPC400170-1GEA 337 EUR
APOSPC400170-1G APOSPC400170-250MG
(3-Chloro-5-fluoropyridin-2-yl)methanamine hydrochloride ≥95%
(3-Chloro-5-fluoropyridin-2-yl)methanamine hydrochloride
Formel: C₆H₆ClFN₂·ClH
Molecular Weight: 197.038 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22689793
CAS-Nummer: 1416714-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR