6-Bromo-5-(trifluoromethoxy)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC400173-1G PC400173-250MG
APOSPC400173-1GEA 510 EUR
APOSPC400173-1G APOSPC400173-250MG
6-Bromo-5-(trifluoromethoxy)-1H-indazole
6-Bromo-5-(trifluoromethoxy)-1H-indazole
Formel: C₈H₄BrF₃N₂O
Molecular Weight: 281,029 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22380592
CAS-Nummer: 1374651-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR