(6-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(Trifluoromethyl)pyridine-2-methylamine hydrochloride

PC400178-1G PC400178-250MG
APOSPC400178-1GEA 393 EUR
APOSPC400178-1G APOSPC400178-250MG
(6-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride
(6-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride
Formel: C₇H₇F₃N₂·ClH
Molecular Weight: 212.6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04117755
CAS-Nummer: 1185022-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR