(2-Fluoro-3-methylpyridin-4-yl)boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-3-methylpyridine-4-boronic acid

PC400180-1G PC400180-250MG PC400180-100MG
APOSPC400180-1GEA 364 EUR
APOSPC400180-1G APOSPC400180-250MG APOSPC400180-100MG
(2-Fluoro-3-methylpyridin-4-yl)boronic acid
(2-Fluoro-3-methylpyridin-4-yl)boronic acid
Formel: C₆H₇BFNO₂
Molecular Weight: 154,935 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16611287
CAS-Nummer: 1451391-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR