4-Chloro-6-(trifluoromethyl)pyridine-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

PC400183-1G PC400183-250MG
APOSPC400183-1GEA 638 EUR
APOSPC400183-1G APOSPC400183-250MG
4-Chloro-6-(trifluoromethyl)pyridine-3-carbonitrile
4-Chloro-6-(trifluoromethyl)pyridine-3-carbonitrile
Formel: C₇H₂ClF₃N₂
Molecular Weight: 206,552 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28118890
CAS-Nummer: 1807217-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR