(2,6-Difluoro-4-isopropoxyphenyl)boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Difluoro-4-isopropyloxyphenylboronic acid

PC400189-1G PC400189-250MG
APOSPC400189-1GEA 377 EUR
APOSPC400189-1G APOSPC400189-250MG
(2,6-Difluoro-4-isopropoxyphenyl)boronic acid
(2,6-Difluoro-4-isopropoxyphenyl)boronic acid
Formel: C₉H₁₁BF₂O₃
Molecular Weight: 215.99 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22689791
CAS-Nummer: 2096337-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR