2-Chloro-5-fluoro-4-(methylthio)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-5-fluoro-4-(methylthio)-pyridine
PC400190-1G
PC400190-250MG
APOSPC400190-1GEA
522
EUR
APOSPC400190-1G
APOSPC400190-250MG
2-Chloro-5-fluoro-4-(methylthio)pyridine
2-Chloro-5-fluoro-4-(methylthio)pyridine
Formel:
C₆H₅ClFNS Molecular Weight: 177,627 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26127744 CAS-Nummer: 1820741-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...